MilliporeSigma
  • Improving device efficiency of polymer/fullerene bulk heterojunction solar cells through enhanced crystallinity and reduced grain boundaries induced by solvent additives.
Improving device efficiency of polymer/fullerene bulk heterojunction solar cells through enhanced crystallinity and reduced grain boundaries induced by solvent additives.
Advanced materials (Deerfield Beach, Fla.) (2011-06-15)
Ming-Shin Su, Chih-Yin Kuo, Mao-Chuan Yuan, U-Ser Jeng, Chun-Jen Su, Kung-Hwa Wei
Sigma-Aldrich
1,6-Diiodohexane, 97%, contains copper as stabilizer