MilliporeSigma
Starch digestion and absorption in nonruminants.
The Journal of nutrition (1992-01-01)
G M Gray
ABSTRACT
Starch digestion and absorption is augmented appreciably by physical processing of grain or legume and by heating to 100 degrees C for several minutes before its ingestion. Starch, a polysaccharide composed of alpha 1,4-linked glucose units (amylose) and alpha 1,4-1,6-linked branched structure (amylopectin), is cleaved in the duodenal cavity by secreted pancreatic alpha-amylase to a disaccharide (maltose), trisaccharide (maltotriose), and branched alpha-dextrins. These final oligosaccharides are hydrolyzed efficiently by complimentary action of three integral brush border enzymes at the intestinal surface: glucoamylase (maltase-glucoamylase, amyloglucosidase), sucrase (maltase-sucrase) and alpha-dextrinase (isomaltase). The final monosaccharide glucose product is then cotransported into the enterocyte along with Na+ by a specific brush border 75-kDa transport protein in the rate-limiting step for overall starch assimilation. By virtue of this sequential luminal and membrane digestion followed by glucose transport, starch is assimilated in a very efficient manner in nonruminants.