MilliporeSigma
  • Surface-enhanced Raman scattering and density functional theoretical study of anthranil adsorbed on colloidal silver particles.
Surface-enhanced Raman scattering and density functional theoretical study of anthranil adsorbed on colloidal silver particles.
The Journal of Physical Chemistry B (2004)
Baia, M, et al.