MilliporeSigma
  • Investigation of selenium speciation in in vitro gastrointestinal extracts of cooked cod by high-performance liquid chromatography-inductively coupled plasma mass spectrometry and electrospray mass spectrometry.
Investigation of selenium speciation in <I>in vitro</I> gastrointestinal extracts of cooked cod by high-performance liquid chromatography-inductively coupled plasma mass spectrometry and electrospray mass spectrometry.
Journal of Analytical Atomic Spectrometry (1177)
JohnaLewis, D.