MilliporeSigma
  • Estimation of ruminal degradation and intestinal digestion of tropical protein resources using the nylon bag technique and the three-step in vitro procedure in dairy cattle on rice straw diets.
Estimation of ruminal degradation and intestinal digestion of tropical protein resources using the nylon bag technique and the three-step <I>in vitro</I> procedure in dairy cattle on rice straw diets.
Asian-Australasian Journal of Animal Sciences (2007)
Promkot, C., M. Wanapat, and P. Rowlinson