MilliporeSigma
  • Lhx6 delineates a pathway mediating innate reproductive behaviors from the amygdala to the hypothalamus.
Lhx6 delineates a pathway mediating innate reproductive behaviors from the amygdala to the hypothalamus.
Neuron (2005-06-10)
Gloria B Choi, Hong-Wei Dong, Andrew J Murphy, David M Valenzuela, George D Yancopoulos, Larry W Swanson, David J Anderson
ABSTRACT
In mammals, innate reproductive and defensive behaviors are mediated by anatomically segregated connections between the amygdala and hypothalamus. This anatomic segregation poses the problem of how the brain integrates activity in these circuits when faced with conflicting stimuli eliciting such mutually exclusive behaviors. Using genetically encoded and conventional axonal tracers, we have found that the transcription factor Lhx6 delineates the reproductive branch of this pathway. Other Lhx proteins mark neurons in amygdalar nuclei implicated in defense. We have traced parallel projections from the posterior medial amygdala, activated by reproductive or defensive olfactory stimuli, respectively, to a point of convergence in the ventromedial hypothalamus. The opposite neurotransmitter phenotypes of these convergent projections suggest a "gate control" mechanism for the inhibition of reproductive behaviors by threatening stimuli. Our data therefore identify a potential neural substrate for integrating the influences of conflicting behavioral cues and a transcription factor family that may contribute to the development of this substrate.