MilliporeSigma
  • Three-component coupling using arynes and DMF: straightforward access to coumarins via ortho-quinone methides.
Three-component coupling using arynes and DMF: straightforward access to coumarins via ortho-quinone methides.
Chemical communications (Cambridge, England) (2011-05-25)
Hiroto Yoshida, Yu Ito, Joji Ohshita
ABSTRACT
ortho-Quinone methides, arising from a formal [2+2] cycloaddition between arynes and DMF, were found to facilely undergo a [4+2] cycloaddition with ester enolates or ketenimine anions to produce diverse coumarins in a straightforward manner.