MilliporeSigma
  • Synthesis of diaryl ethers using an easy-to-prepare, air-stable, soluble copper (I) catalyst.
Synthesis of diaryl ethers using an easy-to-prepare, air-stable, soluble copper (I) catalyst.
Synthetic Communications (2001)
Gujadhur R and Venkataraman D.