MilliporeSigma
  • Assessing arsenic intake from groundwater and rice by residents in Prey Veng province, Cambodia.
Assessing arsenic intake from groundwater and rice by residents in Prey Veng province, Cambodia.
Environmental pollution (Barking, Essex : 1987) (2013-11-16)
Kongkea Phan, Samrach Phan, Savoeun Heng, Laingshun Huoy, Kyoung-Woong Kim
ABSTRACT
We investigated total daily intake of As by residents in Prey Veng province in the Mekong River basin of Cambodia. Groundwater (n = 11), rice (n = 11) and fingernail (n = 23) samples were randomly collected from the households and analyzed for total As by inductively coupled plasma mass spectrometry. Calculation indicated that daily dose of inorganic As was greater than the lower limits on the benchmark dose for a 0.5% increased incidence of lung cancer (BMDL0.5 equals to 3.0 μg d(-1) kg(-1)body wt.). Moreover, positive correlation between As in fingernail and daily dose of As from groundwater and rice and total daily dose of As were found. These results suggest that the Prey Veng residents are exposed to As in groundwater. As in rice is an additional source which is attributable to high As accumulation in human bodies in the Mekong River basin of Cambodia.
Product Description
Rice flour (Total As and As species), ERM®, certified reference material