MilliporeSigma
  • Determination of arsenic species in rice from Thailand and other Asian countries using simple extraction and HPLC-ICP-MS analysis.
Determination of arsenic species in rice from Thailand and other Asian countries using simple extraction and HPLC-ICP-MS analysis.
Journal of agricultural and food chemistry (2013-07-19)
Sumontha Nookabkaew, Nuchanart Rangkadilok, Chulabhorn Mahidol, Gunlatida Promsuk, Jutamaad Satayavivad
ABSTRACT
Human exposure to arsenic (As) via rice consumption is of increasing concern. In the present study, the extraction and HPLC-ICP-MS analysis for As speciation in rice were investigated. A simple extraction with water and digestion with α-amylase followed by the analysis using ion-paring mode HPLC separation was developed. The method showed good extraction efficiencies (generally >80%) and column efficiencies (>90%) for rice samples. The optimization of mobile phase showed well separated peaks of As species. The limits of quantification (LOQ) of As(III), As(V), MMA, and DMA that were calculated based on sample mass were 1.6, 2.0, 2.0, and 1.6 μg kg(-1), respectively. A total of 185 rice samples (various types of rice) collected from different four regions in Thailand and some other Asian countries were analyzed. The total As and inorganic As in rice samples were in the ranges of 22.51-375.39 and 13.89-232.62 μg kg(-1), respectively. The estimated weekly intake of inorganic As from rice by Thai people accounted for 13.91-29.22% of the provisional tolerable weekly intake (PTWI).
Product Description
Rice flour (Total As and As species), ERM®, certified reference material