MilliporeSigma
  • Efficient cyanation of aryl bromides with K4[Fe(CN)6] catalyzed by a palladium-indolylphosphine complex.
Efficient cyanation of aryl bromides with K<SUB>4</SUB>[Fe(CN)<SUB>6</SUB>] catalyzed by a palladium-indolylphosphine complex.
Tetrahedron Letters (2011)
Yeung PY, et al.