MilliporeSigma
  • Assay for both ascorbic and dehydroascorbic acid in dairy foods by high-performance liquid chromatography using precolumn derivatization with methoxy- and ethoxy-1,2-phenylenediamine.
Assay for both ascorbic and dehydroascorbic acid in dairy foods by high-performance liquid chromatography using precolumn derivatization with methoxy- and ethoxy-1,2-phenylenediamine.
Journal of chromatography (1991-05-10)
N Bilic
ABSTRACT
A procedure for the simultaneous determination of both ascorbic and dehydroascorbic acid in dairy foods by high-performance liquid chromatography using precolumn derivatization with 4-methoxy- and 4-ethoxy-1,2-phenylenediamine is presented. The derivatives are isolated by solid-phase extraction and analysed by fluorescence detection on a resin-type reversed-phase column at pH 9. Retention times are 2 and 3.2 min for the derivatives of ascorbic and dehydroascorbic acid, respectively. Relative standard deviations of the within- and between-assay tests are 7.1 and 5.5%, respectively, for ascorbic and 11 and 9%, respectively, dehydroascorbic acid. The limits of detection are 50 and 70 fmol per 5-microl injection for ascorbic and dehydroascorbic acid, respectively.