MilliporeSigma
  • Adsorption of sulfite oxidase on self-assembled monolayers from molecular dynamics simulations.
Adsorption of sulfite oxidase on self-assembled monolayers from molecular dynamics simulations.
Langmuir : the ACS journal of surfaces and colloids (2012-03-06)
Tillmann Utesch, Murat Sezer, Inez M Weidinger, Maria Andrea Mroginski
ABSTRACT
Sulfite oxidase (SO) is an enzyme catalyzing the terminal step of the metabolism of sulfur-containing amino acids that is essential for almost all living organisms. The catalytic activity of SO in vertebrates strongly depends on the efficiency of the intramolecular electron transfer (IET) between the catalytic Moco domain and the cytochrome b5 (cyt b5) domain. The IET process is assumed to be mediated by large domain motions of the cyt b5 domains within the enzyme. Thus, the interaction of SO with charged surfaces may affect the mobility of the cyt b5 domain required for IET and consequently hinder SO activation. In this study, we present a molecular dynamics approach to investigating the ionic strength dependence of the initial surface adsorption of SO in two different conformations-the crystallographic structure and the model structure for an activated SO-onto mixed amino- and hydroxyl-terminated SAMs. The results show for both conformations at low ionic strengths a strong adsorption of the cyt b5 units onto the SAM, which inhibits the domain motion event required for IET. Under higher ion concentrations, however, the interaction with the surface is weakened by the negatively charged ions acting as a buffer and competing in adsorption with the cathodic cyt b5 domains. This competition prevents the immobilization of the cytochrome b5 units onto the surface, allowing the intramolecular domain motions favoring IET. Our predictions support the interpretation of recent experimental spectroelectrochemical studies on SO.
Sigma-Aldrich
8-Amino-1-octanethiol hydrochloride, 95%
Pricing and availability is not currently available.