MilliporeSigma
  • The 'true' L-xylulose reductase of filamentous fungi identified in Aspergillus niger.
The 'true' L-xylulose reductase of filamentous fungi identified in Aspergillus niger.
FEBS letters (2010-07-27)
Dominik Mojzita, Kiira Vuoristo, Outi M Koivistoinen, Merja Penttilä, Peter Richard
ABSTRACT
L-Xylulose reductase is part of the eukaryotic pathway for l-arabinose catabolism. A previously identified L-xylulose reductase in Hypocrea jecorina turned out to be not the 'true' one since it was not upregulated during growth on L-arabinose and the deletion strain showed no reduced L-xylulose reductase activity but instead lost the D-mannitol dehydrogenase activity. In this communication we identified the 'TRUE'L-xylulose reductase in Aspergillus niger. The gene, lxrA (JGI177736), is upregulated on L-arabinose and the deletion results in a strain lacking the NADPH-specific L-xylulose reductase activity and having reduced growth on l-arabinose. The purified enzyme had a K(m) for L-xylulose of 25 mM and a nu(max) of 650 U/mg.