MilliporeSigma
  • Slowly progressive cholangiofibrosis induced in rats by α-naphthylisothiocyanate (ANIT), with particular references to characteristics of macrophages and myofibroblasts.
Slowly progressive cholangiofibrosis induced in rats by α-naphthylisothiocyanate (ANIT), with particular references to characteristics of macrophages and myofibroblasts.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2013-01-10)
Hossain M Golbar, Takeshi Izawa, Chisa Ichikawa, Miyuu Tanaka, Vetnizah Juniantito, Osamu Sawamoto, Mitsuru Kuwamura, Jyoji Yamate
ABSTRACT
A progressive cholangiofibrosis was developed as an animal model in 6-week-old male F344 rats by repeated intraperitoneal injections of α-naphthylisothiocyanate (ANIT) for 19 weeks; liver samples were examined at post-first injection (PFI) weeks 3, 7, 10, 13, 16 and 19, focusing on characteristics of macrophages and myofibroblasts by immunohistochemical analyses. In the affected Glisson's sheath consisting of inflammatory cell infiltrates, bile duct proliferation and advancing fibrosis, the number of macrophages reacting to OX6 (recognizing MHC class II) increased consistently (PFI weeks 3-19), suggesting a central role of antigen presenting cells in the biliary fibrosis; macrophages reacting to ED1 (CD68, reflecting phagocytic activity) and ED2 (CD163, relating to proinflammatory factor production) showed a significantly increased number at PFI weeks 7-19 and PFI weeks 13-19, respectively. Interestingly, macrophages positive for SRA-E5 (CD204, reflecting lipid metabolism) increased at PFI weeks 7-19, and the appearance was limited in the sinusoids around the affected Glisson's sheath. Myofibroblasts appearing in the affected Glisson's sheath reacted to vimentin and desmin at early (PFI weeks 3-7) and mid (PFI weeks 10-13) stages, and then they came to strongly express α-smooth muscle actin at late stage (PFI weeks 16-19). This study shows that macrophages exhibit heterogeneous properties depending on stages and locations; in association with such macrophage populations, myofibroblasts expressing various cytoskeletons participate in cholangiofibrosis. These characteristics would be useful in evaluating the pathogenesis of possible cholangio-toxicants.
Sigma-Aldrich
1-Naphthyl isothiocyanate, 95%