MilliporeSigma
  • A commentary on: Overexpression of Dyrk1A inhibits choline acetyltransferase induction by oleic acid in cellular models of Down syndrome.
A commentary on: Overexpression of Dyrk1A inhibits choline acetyltransferase induction by oleic acid in cellular models of Down syndrome.
Experimental neurology (2013-04-11)
Mara Dierssen