MilliporeSigma
  • Resistance to essential oils affects survival of Salmonella enterica serovars in growing and harvested basil.
Resistance to essential oils affects survival of Salmonella enterica serovars in growing and harvested basil.
Environmental microbiology (2013-05-08)
Guy Kisluk, Emmanuel Kalily, Sima Yaron
ABSTRACT
The number of outbreaks of food-borne illness associated with consumption of fresh products has increased. A recent and noteworthy outbreak occurred in 2007. Basil contaminated with Salmonella enterica serovar Senftenberg was the source of this outbreak. Since basil produces high levels of antibacterial compounds the aim of this study was to investigate if the emerging outbreak reflects ecological changes that occurred as a result of development of resistance to ingredients of the basil oil. We irrigated basil plants with contaminated water containing two Salmonella serovars, Typhimurium and Senftenberg, and showed that Salmonella can survive on the basil plants for at least 100 days. S. Senftenberg counts in the phyllosphere were significantly higher than S. Typhimurium, moreover, S. Senftenberg was able to grow on stored harvested basil leaves. Susceptibility experiments demonstrated that S. Senftenberg is more resistant to basil oil and to its antimicrobial constituents: linalool, estragole and eugenol. This may indicate that S. Senftenberg had adapted to the basil environment by developing resistance to the basil oil. The emergence of resistant pathogens has a significant potential to change the ecology, and opens the way for pathogens to survive in new niches in the environment such as basil and other plants.