MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficacy and safety of the farnesoid X receptor agonist obeticholic acid in patients with type 2 diabetes and nonalcoholic fatty liver disease.
Efficacy and safety of the farnesoid X receptor agonist obeticholic acid in patients with type 2 diabetes and nonalcoholic fatty liver disease.
Gastroenterology (2013-06-04)
Sunder Mudaliar, Robert R Henry, Arun J Sanyal, Linda Morrow, Hanns-Ulrich Marschall, Mark Kipnes, Luciano Adorini, Cathi I Sciacca, Paul Clopton, Erin Castelloe, Paul Dillon, Mark Pruzanski, David Shapiro
ABSTRACT
Obeticholic acid (OCA; INT-747, 6α-ethyl-chenodeoxycholic acid) is a semisynthetic derivative of the primary human bile acid chenodeoxycholic acid, the natural agonist of the farnesoid X receptor, which is a nuclear hormone receptor that regulates glucose and lipid metabolism. In animal models, OCA decreases insulin resistance and hepatic steatosis. We performed a double-blind, placebo-controlled, proof-of-concept study to evaluate the effects of OCA on insulin sensitivity in patients with nonalcoholic fatty liver disease and type 2 diabetes mellitus. Patients were randomly assigned to groups given placebo (n = 23), 25 mg OCA (n = 20), or 50 mg OCA (n = 21) once daily for 6 weeks. A 2-stage hyperinsulinemic-euglycemic insulin clamp was used to measure insulin sensitivity before and after the 6-week treatment period. We also measured levels of liver enzymes, lipid analytes, fibroblast growth factor 19, 7α-hydroxy-4-cholesten-3-one (a BA precursor), endogenous bile acids, and markers of liver fibrosis. When patients were given a low-dose insulin infusion, insulin sensitivity increased by 28.0% from baseline in the group treated with 25 mg OCA (P = .019) and 20.1% from baseline in the group treated with 50 mg OCA (P = .060). Insulin sensitivity increased by 24.5% (P = .011) in combined OCA groups, whereas it decreased by 5.5% in the placebo group. A similar pattern was observed in patients given a high-dose insulin infusion. The OCA groups had significant reductions in levels of γ-glutamyltransferase and alanine aminotransferase and dose-related weight loss. They also had increased serum levels of low-density lipoprotein cholesterol and fibroblast growth factor 19, associated with decreased levels of 7α-hydroxy-4-cholesten-3-one and endogenous bile acids, indicating activation of farnesoid X receptor. Markers of liver fibrosis decreased significantly in the group treated with 25 mg OCA. Adverse experiences were similar among groups. In this phase 2 trial, administration of 25 or 50 mg OCA for 6 weeks was well tolerated, increased insulin sensitivity, and reduced markers of liver inflammation and fibrosis in patients with type 2 diabetes mellitus and nonalcoholic fatty liver disease. Longer and larger studies are warranted. ClinicalTrials.gov, Number: NCT00501592.
Sigma-Aldrich
Chenodeoxycholic acid
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard