MilliporeSigma
  • Effect of citric acid deamidation on in vitro digestibility and antioxidant properties of wheat gluten.
Effect of citric acid deamidation on in vitro digestibility and antioxidant properties of wheat gluten.
Food chemistry (2013-07-23)
Chaoying Qiu, Weizheng Sun, Chun Cui, Mouming Zhao
ABSTRACT
The effects of citric acid deamidation on the physiochemical properties of wheat gluten were investigated. In vitro digestion was carried out to determine changes of molecular weight distribution, amino acids composition and antioxidant efficacy of wheat gluten hydrolysates. Results indicated that citric acid deamidation significantly increased gluten solubility and surface hydrophobicity, at a neutral pH. Deamidation induced molecular weight distribution change of gluten with little proteolysis. Results from FTIR indicated that the α-helix and β-turn of deamidated gluten increased accompanied by a decrease of the β-sheet structure. After deamidation, in vitro pepsin digestibility of wheat gluten decreased, while in vitro pancreatin digestibility increased. The oxygen radical absorbance capacity (ORAC) activity of the in vitro digests decreased with increase of deamidation time. The high Lys and total essential AAs amounts in the final digests suggested that the nutritional values of wheat gluten after deamidation might be enhanced.
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland