MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Preventing oxidation of cellular XRCC1 affects PARP-mediated DNA damage responses.
Preventing oxidation of cellular XRCC1 affects PARP-mediated DNA damage responses.
DNA repair (2013-07-23)
Julie K Horton, Donna F Stefanick, Natalie R Gassman, Jason G Williams, Scott A Gabel, Matthew J Cuneo, Rajendra Prasad, Padmini S Kedar, Eugene F Derose, Esther W Hou, Robert E London, Samuel H Wilson
ABSTRACT
Poly(ADP-ribose) polymerase-1 (PARP-1) binds intermediates of base excision repair (BER) and becomes activated for poly(ADP-ribose) (PAR) synthesis. PAR mediates recruitment and functions of the key BER factors XRCC1 and DNA polymerase β (pol β) that in turn regulate PAR. Yet, the molecular mechanism and implications of coordination between XRCC1 and pol β in regulating the level of PAR are poorly understood. A complex of PARP-1, XRCC1 and pol β is found in vivo, and it is known that pol β and XRCC1 interact through a redox-sensitive binding interface in the N-terminal domain of XRCC1. We confirmed here that both oxidized and reduced forms of XRCC1 are present in mouse fibroblasts. To further understand the importance of the C12-C20 oxidized form of XRCC1 and the interaction with pol β, we characterized cell lines representing stable transfectants in Xrcc1(-/-) mouse fibroblasts of wild-type XRCC1 and two mutants of XRCC1, a novel reduced form with the C12-C20 disulfide bond blocked (C12A) and a reference mutant that is unable to bind pol β (V88R). XRCC1-deficient mouse fibroblasts are extremely hypersensitive to methyl methanesulfonate (MMS), and transfected wild-type and C12A mutant XRCC1 proteins similarly reversed MMS hypersensitivity. However, after MMS exposure the cellular PAR level was found to increase to a much greater extent in cells expressing the C12A mutant than in cells expressing wild-type XRCC1. PARP inhibition resulted in very strong MMS sensitization in cells expressing wild-type XRCC1, but this sensitization was much less in cells expressing the C12A mutant. The results suggest a role for the oxidized form of XRCC1 in the interaction with pol β in (1) controlling the PAR level after MMS exposure and (2) enabling the extreme cytotoxicity of PARP inhibition during the MMS DNA damage response.
Sigma-Aldrich
Methyl methanesulfonate, 99%
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-Amino-1,8-naphthalimide
Sigma-Aldrich
1-Naphthylamine, 97%
Supelco
1-Naphthylamine, analytical standard
Millipore
Nitrate Reagent A, suitable for microbiology