MilliporeSigma
  • Copper-catalyzed nucleophilic trifluoromethylation of propargylic halides.
Copper-catalyzed nucleophilic trifluoromethylation of propargylic halides.
Chemical communications (Cambridge, England) (2013-07-26)
Yoshihiro Miyake, Shin-ichi Ota, Masashi Shibata, Kazunari Nakajima, Yoshiaki Nishibayashi
ABSTRACT
Reactions of propargylic halides with trifluoromethyltrimethylsilane in the presence of a catalytic amount of copper(I) thiophene-2-carboxylate (CuTC) have been found to give the corresponding trifluoromethylated products in good to high yields with a high selectivity.
Sigma-Aldrich
2-Thiophenecarboxylic acid, ReagentPlus®, 99%