MilliporeSigma
  • Deletion of ATG5 shows a role of autophagy in salivary homeostatic control.
Deletion of ATG5 shows a role of autophagy in salivary homeostatic control.
Journal of dental research (2013-07-26)
M Morgan-Bathke, H H Lin, A M Chibly, W Zhang, X Sun, C-H Chen, P Flodby, Z Borok, R Wu, D Arnett, R R Klein, D K Ann, K H Limesand
ABSTRACT
Autophagy is a catabolic pathway utilized to maintain a balance among the synthesis, degradation, and recycling of cellular components, thereby playing a role in cell growth, development, and homeostasis. Previous studies revealed that a conditional knockout of essential member(s) of autophagy in a variety of tissues causes changes in structure and function of these tissues. Acinar cell-specific expression of knocked-in Cre recombinase through control of aquaporin 5 (Aqp5) promoter/enhancer (Aqp5-Cre) allows us to specifically inactivate Atg5, a protein necessary for autophagy, in salivary acinar cells of Atg5(f/f);Aqp5-Cre mice. There was no difference in apoptotic or proliferation levels in salivary glands of Atg5/Cre mice from each genotype. However, H&E staining and electron microscopy studies revealed modestly enlarged acinar cells and accumulated secretory granules in salivary glands of Atg5(f/f);Aqp5-Cre mice. Salivary flow rates and amylase contents of Atg5/Cre mice indicated that acinar-specific inactivation of ATG5 did not alter carbachol-evoked saliva and amylase secretion. Conversely, autophagy intersected with salivary morphological and secretory manifestations induced by isoproterenol administration. These results identified a role for autophagy as a homeostasis control in salivary glands. Collectively, Atg5(f/f);Aqp5-Cre mice would be a useful tool to enhance our understanding of autophagy in adaptive responses following targeted head and neck radiation or Sjögren syndrome.
Sigma-Aldrich
Isoprenaline hydrochloride