MilliporeSigma
  • Simultaneous enhanced diagnosis and photodynamic therapy of photosensitizer-doped perylene nanoparticles via doping, fluorescence resonance energy transfer, and antenna effect.
Simultaneous enhanced diagnosis and photodynamic therapy of photosensitizer-doped perylene nanoparticles via doping, fluorescence resonance energy transfer, and antenna effect.
Chemical communications (Cambridge, England) (2013-08-03)
Jinfeng Zhang, Feifei An, Yanan Li, Caijun Zheng, Yinlong Yang, Xiujuan Zhang, Xiaohong Zhang
ABSTRACT
We demonstrate a new strategy for preparing photosensitizer-doped perylene nanoparticles via doping, fluorescence resonance energy transfer, and antenna effect for simultaneous enhanced diagnosis and photodynamic therapy.
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%