MilliporeSigma
  • Property changes of urinary nanocrystallites and urine of uric acid stone formers after taking potassium citrate.
Property changes of urinary nanocrystallites and urine of uric acid stone formers after taking potassium citrate.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Guang-Na Zhang, Jian-Ming Ouyang, Jun-Fa Xue, Yun-Feng Shang
ABSTRACT
The property changes of urinary nanocrystallites in 20 cases of uric acid (UA) stone formers after 1 week of potassium citrate (K3cit) intake were comparatively studied by X-ray diffraction analysis, Fourier transform infrared spectroscopy, nanoparticle size analysis, and transmission electron microscopy. Before K3cit intake, the urinary crystallites mainly contained UA and calcium oxalate. After K3cit intake, the components changed to urate and UA; the qualities, species, and amounts of aggregated crystallites decreased; urine pH, citrate, and glycosaminoglycan excretions increased; and UA excretion, Zeta potential, and crystallite size decreased. The stability of crystallites followed the order: controls>patients after taking K3cit>patients before taking K3cit. Therefore, the components of urinary stones were closely related to the components of urinary crystallites.
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland