MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Enhanced wear and corrosion in modular tapers in total hip replacement is associated with the contact area and surface topography.
Enhanced wear and corrosion in modular tapers in total hip replacement is associated with the contact area and surface topography.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2013-08-24)
Anna Panagiotidou, Jay Meswania, Jia Hua, Sarah Muirhead-Allwood, Alister Hart, Gordon Blunn
ABSTRACT
Widespread concern exists about adverse tissue reactions after metal-on-metal (MoM) total hip replacement (THR). Concerns have also been expressed with wear and corrosion of taper junctions in THR. We report the effect of surface finish and contact area associated with a single combination of materials of modular tapers. In an in vitro test, we investigated the head/neck (CoCrMo/Ti) interface of modular THRs using commercially available heads. Wear and corrosion of taper surfaces was compared following a 10 million loading cycle. Surface parameters and profiles were measured before and after testing. Electrochemical static and dynamic corrosion tests were performed under loaded and non-loaded conditions. After the load test, the surface roughness parameters on the head taper were significantly increased where the head/neck contact area was reduced. Similarly, the surface roughness parameters on the head taper were significantly increased where rough neck tapers were used. Corrosion testing showed breaching of the passive film on the rough but not the smooth neck tapers. Thus, surface area and surface finish are important factors in wear and corrosion at modular interfaces.
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis