MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Catalytic asymmetric Petasis reactions of vinylboronates.
The Journal of organic chemistry (2013-08-27)
Xianglin Shi, William F Kiesman, Anna Levina, Zhili Xin
ABSTRACT
Binaphthol-catalyzed asymmetric Petasis reactions of salicylaldehydes with dibutyl vinylboronates and secondary amines in the presence of 4 Å molecular sieves (MS) afforded products with up to 99% ee in isolated yields of 39-94%. The 99% ee of the product indicated that the reaction by the binaphthol-catalyzed pathway was roughly 500 times faster than the uncatalyzed pathway. NMR experiments ((1)H and (11)B) showed that the amine component played a role in triggering the reaction between the binaphthol catalyst and the vinylboronate in the catalytic reaction sequence. The 4 Å MS enhanced both the rate and enantioselectivity by effective removal of water from the reaction system. A novel rearrangement reaction of the unconjugated allylic amine Petasis reaction product to a conjugated allylic amine was also observed.
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%