MilliporeSigma
  • Antioxidant and vitamin E transport genes and risk of high-grade prostate cancer and prostate cancer recurrence.
Antioxidant and vitamin E transport genes and risk of high-grade prostate cancer and prostate cancer recurrence.
The Prostate (2013-09-17)
Scott R Bauer, Erin L Richman, Eduardo Sosa, Vivian Weinberg, Xiaoling Song, John S Witte, Peter R Carroll, June M Chan
ABSTRACT
Observational studies suggest an inverse association between vitamin E and risk of prostate cancer, particularly aggressive tumors. However, three large randomized controlled trials have reported conflicting results. Thus, we examined circulating vitamin E and vitamin E-related genes in relation to risk of high-grade prostate cancer and prostate cancer recurrence among men initially diagnosed with clinically organ-confined disease. We measured circulating α- and γ-tocopherol and genotyped 30 SNPs in SOD1, SOD2, SOD3, TTPA, and SEC14L2 among 573 men with organ-confined prostate cancer who underwent radical prostatectomy. We examined associations between circulating vitamin E, genotypes, and risk of high-grade prostate cancer (Gleason grade ≥ 8 or 7 with primary score ≥ 4; n = 117) using logistic regression, and risk of recurrence (56 events; 3.7 years median follow-up) using Cox proportional hazards regression. Circulating γ-tocopherol was associated with an increased risk of high-grade prostate cancer (Q4 v. Q1 odds ratio [OR] = 1.87; 95% confidence intervals [CI]: 0.97-3.58; P trend =0.02). The less common allele in SOD3 rs699473 was associated with an increased risk of high-grade disease (T > C: OR = 1.40, 95% CI: 1.04-1.89). Two independent SNPs in SOD1 were inversely associated with prostate cancer recurrence in additive models (rs17884057 hazard ratio [HR] = 0.49, 95%CI: 0.25-0.96; rs9967983 HR = 0.62, 95% CI: 0.40-0.95). Among men with clinically organ-confined prostate cancer, genetic variation in SOD may be associated with risk of high-grade disease at diagnosis and disease recurrence. Circulating γ-tocopherol levels may also be associated with an increased risk of high-grade disease at diagnosis.
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g