MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Benefits and harms of adhesion barriers for abdominal surgery: a systematic review and meta-analysis.
Benefits and harms of adhesion barriers for abdominal surgery: a systematic review and meta-analysis.
Lancet (London, England) (2013-10-01)
Richard P G Ten Broek, Martijn W J Stommel, Chema Strik, Cornelis J H M van Laarhoven, Frederik Keus, Harry van Goor
ABSTRACT
Formation of adhesions after peritoneal surgery results in high morbidity. Barriers to prevent adhesion are seldom applied, despite their ability to reduce the severity of adhesion formation. We evaluated the benefits and harms of four adhesion barriers that have been approved for clinical use. In this systematic review and meta-analysis, we searched PubMed, CENTRAL, and Embase for randomised clinical trials assessing use of oxidised regenerated cellulose, hyaluronate carboxymethylcellulose, icodextrin, or polyethylene glycol in abdominal surgery. Two researchers independently identified reports and extracted data. We compared use of a barrier with no barrier for nine predefined outcomes, graded for clinical relevance. The primary outcome was reoperation for adhesive small bowel obstruction. We assessed systematic error, random error, and design error with the error matrix approach. This study is registered with PROSPERO, number CRD42012003321. Our search returned 1840 results, from which 28 trials (5191 patients) were included in our meta-analysis. The risks of systematic and random errors were low. No trials reported data for the effect of oxidised regenerated cellulose or polyethylene glycol on reoperations for adhesive small bowel obstruction. Oxidised regenerated cellulose reduced the incidence of adhesions (relative risk [RR] 0·51, 95% CI 0·31-0·86). Some evidence suggests that hyaluronate carboxymethylcellulose reduces the incidence of reoperations for adhesive small bowel obstruction (RR 0·49, 95% CI 0·28-0·88). For icodextrin, reoperation for adhesive small bowel obstruction did not differ significantly between groups (RR 0·33, 95% CI 0·03-3·11). No barriers were associated with an increase in serious adverse events. Oxidised regenerated cellulose and hyaluronate carboxymethylcellulose can safely reduce clinically relevant consequences of adhesions. None.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, SAJ first grade
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP