MilliporeSigma
  • Design, synthesis, and biological evaluation of 14-heteroaromatic-substituted naltrexone derivatives: pharmacological profile switch from mu opioid receptor selectivity to mu/kappa opioid receptor dual selectivity.
Design, synthesis, and biological evaluation of 14-heteroaromatic-substituted naltrexone derivatives: pharmacological profile switch from mu opioid receptor selectivity to mu/kappa opioid receptor dual selectivity.
Journal of medicinal chemistry (2013-10-23)
Yunyun Yuan, Saheem A Zaidi, Orgil Elbegdorj, Lindsey C K Aschenbach, Guo Li, David L Stevens, Krista L Scoggins, William L Dewey, Dana E Selley, Yan Zhang
ABSTRACT
On the basis of a mu opioid receptor (MOR) homology model and the isosterism concept, three generations of 14-heteroaromatically substituted naltrexone derivatives were designed, synthesized, and evaluated as potential MOR-selective ligands. The first-generation ligands appeared to be MOR-selective, whereas the second and the third generation ones showed MOR/kappa opioid receptor (KOR) dual selectivity. Docking of ligands 2 (MOR selective) and 10 (MOR/KOR dual selective) to the three opioid receptor crystal structures revealed a nonconserved-residue-facilitated hydrogen-bonding network that could be responsible for their distinctive selectivity profiles. The MOR/KOR dual-selective ligand 10 showed no agonism and acted as a potent antagonist in the tail-flick assay. It also produced less severe opioid withdrawal symptoms than naloxone in morphine-dependent mice. In conclusion, ligand 10 may serve as a novel lead compound to develop MOR/KOR dual-selective ligands, which might possess unique therapeutic value for opioid addiction treatment.
Product Description
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder