MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Femoral and tibial component rotation in total knee arthroplasty: methods and consequences.
Femoral and tibial component rotation in total knee arthroplasty: methods and consequences.
The bone & joint journal (2013-11-06)
R D Scott
ABSTRACT
At least four ways have been described to determine femoral component rotation, and three ways to determine tibial component rotation in total knee replacement (TKR). Each method has its advocates and each has an influence on knee kinematics and the ultimate short and long term success of TKR. Of the four femoral component methods, the author prefers rotating the femoral component in flexion to that amount that establishes a stable symmetrical flexion gap. This judgement is made after the soft tissues of the knee have been balanced in extension. Of the three tibial component methods, the author prefers rotating the tibial component into congruency with the established femoral component rotation with the knee is in extension. This yields a rotationally congruent articulation during weight-bearing and should minimise the torsional forces being transferred through a conforming tibial insert, which could lead to wear to the underside of the tibial polyethylene. Rotating platform components will compensate for any mal-rotation, but can still lead to pain if excessive tibial insert rotation causes soft-tissue impingement.
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material