- Field evaluation and user acceptability of repellent formulations containing DEET against mosquitoes in Australia.
Field evaluation and user acceptability of repellent formulations containing DEET against mosquitoes in Australia.
Journal of the American Mosquito Control Association (2013-11-10)
Stephen P Frances
PMID24199505
ABSTRACT
Field efficacy trials comparing 2 formulations of deet against mosquitoes in Redcliffe, Queensland, Australia were conducted in February 2009. A formulation containing 35% deet in a gel (Australian Defence Force deet) provided > 95% protection for 3 h, while a formulation containing 40% deet in ethanol (Bushman) in a spray applicator provided > 95% for 6 h. A user acceptability study showed that 82% of soldiers using the Bushman formulation during contingency operations for 14-28 days in Timor-Leste would recommend this formulation to others and believed that the formulation provided protection against mosquitoes.