MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Preparation of polyvinylidene fluoride nanofiber membrane and its antibacterial characteristics with nanosilver or graphene oxide.
Preparation of polyvinylidene fluoride nanofiber membrane and its antibacterial characteristics with nanosilver or graphene oxide.
Journal of nanoscience and nanotechnology (2013-11-12)
Byungpyo Hong, Hyemin Jung, Hongsik Byun
ABSTRACT
Polyvinylidene fluoride (PVdF) (Kynar 761) nanofibers were prepared by electrospinning at an external voltage of 6-10 kV, a traveling distance of 7-15 cm and flow rate of 0.4-1 ml/hr. Although the diameter of the fiber was not significantly changed, the electrospinning conditions affected the overall distribution of diameter. This is probably due to the interactions, both attraction and repulsion, of positive charges on polymer solutions and the electrically grounded collector. Especially, the effect of voltage on the distribution of diameter was investigated in this study. The final PVdF nanofiber membrane showed narrow pore-size distribution and high water flux compared with the commercial MF membrane. PVdF nanofiber membranes incorporated nanosilver or graphene oxide were also prepared as nanosilver and graphene have an antibacterial activity. It was found that more than 200 ppm of silver nanoparticles in the PVdF nanofiber had 99.9% of growth inhibition of Staphylococcus aureus and Klebsiella pneumonia. It was also found that 0.2 wt% of graphene oxide in the PVdF electrospinning solution had 99.6% of disinfection property to E-Coli.
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Poly(vinylidene fluoride), beads or pellets
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis