MilliporeSigma
  • Brostallicin versus doxorubicin as first-line chemotherapy in patients with advanced or metastatic soft tissue sarcoma: an European Organisation for Research and Treatment of Cancer Soft Tissue and Bone Sarcoma Group randomised phase II and pharmacogenetic study.
Brostallicin versus doxorubicin as first-line chemotherapy in patients with advanced or metastatic soft tissue sarcoma: an European Organisation for Research and Treatment of Cancer Soft Tissue and Bone Sarcoma Group randomised phase II and pharmacogenetic study.
European journal of cancer (Oxford, England : 1990) (2013-11-13)
H Gelderblom, J Y Blay, B M Seddon, M Leahy, I Ray-Coquard, S Sleijfer, J M Kerst, P Rutkowski, S Bauer, M Ouali, S Marreaud, R J H M van der Straaten, H-J Guchelaar, S D Weitman, P C W Hogendoorn, P Hohenberger
ABSTRACT
Brostallicin is a DNA minor groove binder that has shown activity in patients with soft tissue sarcoma (STS) failing first-line therapy. The present study assessed the safety and efficacy of first-line brostallicin in patients with advanced or metastatic STS >60 years or not fit enough to receive combination chemotherapy. A prospective explorative pharmacogenetic analysis was undertaken in parallel. Patients were randomised in a 2:1 ratio between IV brostallicin 10mg/m(2) and doxorubicin 75 mg/m(2) once every 3 weeks for a maximum of six cycles. Disease stabilisation at 26 weeks (primary end-point) was considered a 'success'. Further testing of brostallicin was warranted if ≥ 35 'successes' were observed in the first 72 eligible patients treated with brostallicin. In addition, patients were genotyped for glutathione S transferase (GST) polymorphisms. One hundred and eighteen patients were included (79 brostallicin and 39 doxorubicin). Brostallicin was well tolerated in comparison to doxorubicin with less grade 3-4 neutropenia (67% versus 95%), grade 2-3 systolic dysfunction (0% versus 11%), alopecia (17% versus 61%) and grade 2-3 mucositis (0% versus 18%). For brostallicin versus doxorubicin, 'successes' were observed in 5/77 versus 10/36, progression free survival at 1 year was 6.5% versus 15.6%, objective response rate was 3.9% versus 22.2% and overall survival at 1 year was 50.5% versus 57.9%, respectively. Only GSTA1 genotype was significantly associated with success rate of doxorubicin treatment. Brostallicin cannot be recommended at this dose and schedule in this patient population as first-line therapy. GSTA1 genotype may be predictive for doxorubicin efficacy but warrants further study.
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein