MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Coordinated expression of fdxD and molybdenum nitrogenase genes promotes nitrogen fixation by Rhodobacter capsulatus in the presence of oxygen.
Coordinated expression of fdxD and molybdenum nitrogenase genes promotes nitrogen fixation by Rhodobacter capsulatus in the presence of oxygen.
Journal of bacteriology (2013-11-26)
Marie-Christine Hoffmann, Alexandra Müller, Maria Fehringer, Yvonne Pfänder, Franz Narberhaus, Bernd Masepohl
ABSTRACT
Rhodobacter capsulatus is able to grow with N2 as the sole nitrogen source using either a molybdenum-dependent or a molybdenum-free iron-only nitrogenase whose expression is strictly inhibited by ammonium. Disruption of the fdxD gene, which is located directly upstream of the Mo-nitrogenase genes, nifHDK, abolished diazotrophic growth via Mo-nitrogenase at oxygen concentrations still tolerated by the wild type, thus demonstrating the importance of FdxD under semiaerobic conditions. In contrast, FdxD was not beneficial for diazotrophic growth depending on Fe-nitrogenase. These findings suggest that the 2Fe2S ferredoxin FdxD specifically supports the Mo-nitrogenase system, probably by protecting Mo-nitrogenase against oxygen, as previously shown for its Azotobacter vinelandii counterpart, FeSII. Expression of fdxD occurred under nitrogen-fixing conditions, but not in the presence of ammonium. Expression of fdxD strictly required NifA1 and NifA2, the transcriptional activators of the Mo-nitrogenase genes, but not AnfA, the transcriptional activator of the Fe-nitrogenase genes. Expression of the fdxD and nifH genes, as well as the FdxD and NifH protein levels, increased with increasing molybdate concentrations. Molybdate induction of fdxD was independent of the molybdate-sensing regulators MopA and MopB, which repress anfA transcription at micromolar molybdate concentrations. In this report, we demonstrate the physiological relevance of an fesII-like gene, fdxD, and show that the cellular nitrogen and molybdenum statuses are integrated to control its expression.
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis