MilliporeSigma
69-year-old woman with ascites, hypoxia, and weight loss.
Mayo Clinic proceedings (2013-12-03)
Megha Prasad, Michael E Wilson, Michael D McGoon
Product Description
Supelco
Sildenafil solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®