MilliporeSigma
  • Angiotensin II-dependent TGF-β signaling contributes to Loeys-Dietz syndrome vascular pathogenesis.
Angiotensin II-dependent TGF-β signaling contributes to Loeys-Dietz syndrome vascular pathogenesis.
The Journal of clinical investigation (2013-12-21)
Elena M Gallo, David C Loch, Jennifer P Habashi, Juan F Calderon, Yichun Chen, Djahida Bedja, Christel van Erp, Elizabeth E Gerber, Sarah J Parker, Kimberly Sauls, Daniel P Judge, Sara K Cooke, Mark E Lindsay, Rosanne Rouf, Loretha Myers, Colette M ap Rhys, Kathleen C Kent, Russell A Norris, David L Huso, Harry C Dietz
ABSTRACT
Loeys-Dietz syndrome (LDS) is a connective tissue disorder that is characterized by a high risk for aneurysm and dissection throughout the arterial tree and phenotypically resembles Marfan syndrome. LDS is caused by heterozygous missense mutations in either TGF-β receptor gene (TGFBR1 or TGFBR2), which are predicted to result in diminished TGF-β signaling; however, aortic surgical samples from patients show evidence of paradoxically increased TGF-β signaling. We generated 2 knockin mouse strains with LDS mutations in either Tgfbr1 or Tgfbr2 and a transgenic mouse overexpressing mutant Tgfbr2. Knockin and transgenic mice, but not haploinsufficient animals, recapitulated the LDS phenotype. While heterozygous mutant cells had diminished signaling in response to exogenous TGF-β in vitro, they maintained normal levels of Smad2 phosphorylation under steady-state culture conditions, suggesting a chronic compensation. Analysis of TGF-β signaling in the aortic wall in vivo revealed progressive upregulation of Smad2 phosphorylation and TGF-β target gene output, which paralleled worsening of aneurysm pathology and coincided with upregulation of TGF-β1 ligand expression. Importantly, suppression of Smad2 phosphorylation and TGF-β1 expression correlated with the therapeutic efficacy of the angiotensin II type 1 receptor antagonist losartan. Together, these data suggest that increased TGF-β signaling contributes to postnatal aneurysm progression in LDS.
Product Description
Losartan potassium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Supelco
Losartan potassium
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Anti-phospho-Smad2 (Ser465/467) Antibody, clone A5S, rabbit monoclonal, culture supernatant, clone A5S, Upstate®