MilliporeSigma
  • Wound infiltration with local anaesthetic agents for laparoscopic cholecystectomy.
Wound infiltration with local anaesthetic agents for laparoscopic cholecystectomy.
The Cochrane database of systematic reviews (2014-03-13)
Sofronis Loizides, Kurinchi Selvan Gurusamy, Myura Nagendran, Michele Rossi, Gian Piero Guerrini, Brian R Davidson
ABSTRACT
While laparoscopic cholecystectomy is generally considered to be less painful than open surgery, pain is one of the important reasons for delayed discharge after day surgery resulting in overnight stay following laparoscopic cholecystectomy. The safety and effectiveness of local anaesthetic wound infiltration in people undergoing laparoscopic cholecystectomy is not known. To assess the benefits and harms of local anaesthetic wound infiltration in patients undergoing laparoscopic cholecystectomy and to identify the best method of local anaesthetic wound infiltration with regards to the type of local anaesthetic, dosage, and time of administration of the local anaesthetic. We searched the Cochrane Central Register of Controlled Trials (CENTRAL), MEDLINE, EMBASE, and Science Citation Index Expanded until February 2013 to identify studies of relevance to this review. We included randomised clinical trials for benefit and quasi-randomised and comparative non-randomised studies for treatment-related harms. Only randomised clinical trials (irrespective of language, blinding, or publication status) comparing local anaesthetic wound infiltration versus placebo, no intervention, or inactive control during laparoscopic cholecystectomy, trials comparing different local anaesthetic agents for local anaesthetic wound infiltration, and trials comparing the different times of local anaesthetic wound infiltration were considered for the review. Two review authors collected the data independently. We analysed the data with both fixed-effect and random-effects meta-analysis models using RevMan. For each outcome, we calculated the risk ratio (RR) or mean difference (MD) with 95% confidence interval (CI). Twenty-six trials fulfilled the inclusion criteria of the review. All the 26 trials except one trial of 30 participants were at high risk of bias. Nineteen of the trials with 1263 randomised participants provided data for this review. Ten of the 19 trials compared local anaesthetic wound infiltration versus inactive control. One of the 19 trials compared local anaesthetic wound infiltration with two inactive controls, normal saline and no intervention. Two of the 19 trials had four arms comparing local anaesthetic wound infiltration with inactive controls in the presence and absence of co-interventions to decrease pain after laparoscopic cholecystectomy. Four of the 19 trials had three or more arms that could be included for the comparison of local anaesthetic wound infiltration versus inactive control and different methods of local anaesthetic wound infiltration. The remaining two trials compared different methods of local anaesthetic wound infiltration.Most trials included only low anaesthetic risk people undergoing elective laparoscopic cholecystectomy. Seventeen trials randomised a total of 1095 participants to local anaesthetic wound infiltration (587 participants) versus no local anaesthetic wound infiltration (508 participants). Various anaesthetic agents were used but bupivacaine was the commonest local anaesthetic used. There was no mortality in either group in the seven trials that reported mortality (0/280 (0%) in local anaesthetic infiltration group versus 0/259 (0%) in control group). The effect of local anaesthetic on the proportion of people who developed serious adverse events was imprecise and compatible with increase or no difference in serious adverse events (seven trials; 539 participants; 2/280 (0.8%) in local anaesthetic group versus 1/259 (0.4%) in control; RR 2.00; 95% CI 0.19 to 21.59; very low quality evidence). None of the serious adverse events were related to local anaesthetic wound infiltration. None of the trials reported patient quality of life. The proportion of participants who were discharged as day surgery patients was higher in the local anaesthetic infiltration group than in the no local anaesthetic infiltration group (one trial; 97 participants; 33/50 (66.0%) in the local anaesthetic group versus 20/47 (42.6%) in the control group; RR 1.55; 95% CI 1.05 to 2.28; very low quality evidence). The effect of local anaesthetic on the length of hospital stay was compatible with a decrease, increase, or no difference in the length of hospital stay between the two groups (four trials; 327 participants; MD -0.26 days; 95% CI -0.67 to 0.16; very low quality evidence). The pain scores as measured by the visual analogue scale (0 to 10 cm) were lower in the local anaesthetic infiltration group than the control group at 4 to 8 hours (13 trials; 806 participants; MD -1.33 cm on the VAS; 95% CI -1.54 to -1.12; very low quality evidence) and 9 to 24 hours (12 trials; 756 participants; MD -0.36 cm on the VAS; 95% CI -0.53 to -0.20; very low quality evidence). The effect of local anaesthetic on the time taken to return to normal activity between the two groups was imprecise and compatible with a decrease, increase, or no difference in the time taken to return to normal activity (two trials; 195 participants; MD 0.14 days; 95% CI -0.59 to 0.87; very low quality evidence). None of the trials reported on return to work.Four trials randomised a total of 149 participants to local anaesthetic wound infiltration prior to skin incision (74 participants) versus local anaesthetic wound infiltration at the end of surgery (75 participants). Two trials randomised a total of 176 participants to four different local anaesthetics (bupivacaine, levobupivacaine, ropivacaine, neosaxitoxin). Although there were differences between the groups in some outcomes the changes were not consistent. There was no evidence to support the preference of one local anaesthetic over another or to prefer administration of local anaesthetic at a specific time compared with another. Serious adverse events were rare in studies evaluating local anaesthetic wound infiltration (very low quality evidence). There is very low quality evidence that infiltration reduces pain in low anaesthetic risk people undergoing elective laparoscopic cholecystectomy. However, the clinical importance of this reduction in pain is likely to be small. Further randomised clinical trials at low risk of systematic and random errors are necessary. Such trials should include important clinical outcomes such as quality of life and time to return to work in their assessment.