MilliporeSigma
  • Inhibition of Bak activation by VDAC2 is dependent on the Bak transmembrane anchor.
Inhibition of Bak activation by VDAC2 is dependent on the Bak transmembrane anchor.
The Journal of biological chemistry (2010-09-21)
Michael Lazarou, Diana Stojanovski, Ann E Frazier, Aneta Kotevski, Grant Dewson, William J Craigen, Ruth M Kluck, David L Vaux, Michael T Ryan
ABSTRACT
Bax and Bak are pro-apoptotic factors that are required for cell death by the mitochondrial or intrinsic pathway. Bax is found in an inactive state in the cytosol and upon activation is targeted to the mitochondrial outer membrane where it releases cytochrome c and other factors that cause caspase activation. Although Bak functions in the same way as Bax, it is constitutively localized to the mitochondrial outer membrane. In the membrane, Bak activation is inhibited by the voltage-dependent anion channel isoform 2 (VDAC2) by an unknown mechanism. Using blue native gel electrophoresis, we show that in healthy cells endogenous inactive Bak exists in a 400-kDa complex that is dependent on the presence of VDAC2. Activation of Bak is concomitant with its release from the 400-kDa complex and the formation of lower molecular weight species. Furthermore, substitution of the Bak transmembrane anchor with that of the mitochondrial outer membrane tail-anchored protein hFis1 prevents association of Bak with the VDAC2 complex and increases the sensitivity of cells to an apoptotic stimulus. Our results suggest that VDAC2 interacts with the hydrophobic tail of Bak to sequester it in an inactive state in the mitochondrial outer membrane, thereby raising the stimulation threshold necessary for permeabilization of the mitochondrial outer membrane and cell death.
Sigma-Aldrich
Anti-VDAC2 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.