MilliporeSigma
  • Physical heterogeneity of bovine gamma-globulins: characterization of gamma-M and gamma-G globulins.
Physical heterogeneity of bovine gamma-globulins: characterization of gamma-M and gamma-G globulins.
Archives of biochemistry and biophysics (1965-10-01)
F A Murphy, J W Osebold, O Aalund
Sigma-Aldrich
γ-Globulins from bovine blood, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
γ-Globulins from bovine blood, ≥97% (agarose gel electrophoresis)