MilliporeSigma
  • Electrochemical method for the synthesis of disulfides of 2-(benzo[d]thiazol(or oxazol)-2-ylamino)-5-morpholinobenzenethiol.
Electrochemical method for the synthesis of disulfides of 2-(benzo[d]thiazol(or oxazol)-2-ylamino)-5-morpholinobenzenethiol.
The Journal of organic chemistry (2013-05-01)
Roya Esmaili, Davood Nematollahi
ABSTRACT
Electrochemical synthesis of two new disulfides of 2-(benzo[d]thiazol(or oxazol)-2-ylamino)-5-morpholinobenzene thiols was carried out via the electrooxidation of 4-morpholinoaniline in the presence of 2-mercaptobenzothiazole and 2-mercaptobenzoxazole. Our results indicate that electrogenerated p-quinonediimine participated in a Michael-type addition reaction with 2-SH-benzazoles and after intramolecular nucleophilic substitution reaction and electrooxidative disulfide bond formation were converted to the corresponding disulfide compounds.