MilliporeSigma
  • 1H, 13C and 15N nuclear magnetic resonance coordination shifts in Au(III), Pd(II) and Pt(II) chloride complexes with phenylpyridines.
1H, 13C and 15N nuclear magnetic resonance coordination shifts in Au(III), Pd(II) and Pt(II) chloride complexes with phenylpyridines.
Magnetic resonance in chemistry : MRC (2009-05-28)
Leszek Pazderski, Jaromír Tousek, Jerzy Sitkowski, Lech Kozerski, Edward Szłyk
ABSTRACT
1H, 13C and 15N nuclear magnetic resonance studies of gold(III), palladium(II) and platinum(II) chloride complexes with phenylpyridines (PPY: 4-phenylpyridine, 4ppy; 3-phenylpyridine, 3ppy; and 2-phenylpyridine, 2ppy) having the general formulae [Au(PPY)Cl3], trans-/cis-[Pd(PPY)2Cl2] and trans-/cis-[Pt(PPY)2Cl2] were performed and the respective chemical shifts (delta1H, delta13C and delta15N) reported. 1H, 13C and 15N coordination shifts (i.e. differences between chemical shifts of the same atom in the complex and ligand molecules: Delta(coord)(1H) = delta(complex)(1H)-delta(ligand)(1H), Delta(coord)(13C) = delta(complex)(13C)-delta(ligand)(13C), Delta(coord)(15N) = delta(complex)(15N)-delta(ligand)(15N)) were discussed in relation to the type of the central atom (Au(III), Pd(II) and Pt(II)), geometry (trans-/cis-) and the position of a phenyl group in the pyridine ring system.