MilliporeSigma
  • Organic crystal engineering with 1,4-piperazine-2,5-diones. 6. Studies of the hydrogen-bond association of cyclo[(2-methylamino-4,7-dimethoxyindan-2-carboxylic acid)(2-amino-4,7-dimethoxyindan-2-carboxylic acid)].
Organic crystal engineering with 1,4-piperazine-2,5-diones. 6. Studies of the hydrogen-bond association of cyclo[(2-methylamino-4,7-dimethoxyindan-2-carboxylic acid)(2-amino-4,7-dimethoxyindan-2-carboxylic acid)].
The Journal of organic chemistry (2005-10-22)
Robin A Weatherhead-Kloster, Hugh D Selby, Walter B Miller Iii, Eugene A Mash
ABSTRACT
[structure: see text] The title 1,4-piperazine-2,5-dione was synthesized in 23% yield over six steps from ethyl 2-amino-4,7-dimethoxyindan-2-carboxylate. Crystallization by slow diffusion of ether into a chloroform solution and by slow evaporation of an ethanol-chloroform-benzene solution produced polymorphic crystalline forms as determined by single-crystal X-ray analysis. The polymorphs exhibited different hydrogen-bonding networks. The association of this piperazinedione in solution was studied using mass spectrometric and nuclear magnetic resonance spectroscopic techniques. The MS and NMR data were interpreted using the solid-state structures as models for solution aggregation. Association constants extracted from the NMR data are in line with those of other cyclic cis amides in chloroform solvent.