MilliporeSigma
  • Delta agonist hydroxy bioisosteres: the discovery of 3-((1-benzylpiperidin-4-yl){4-[(diethylamino)carbonyl]phenyl}amino)benzamide with improved delta agonist activity and in vitro metabolic stability.
Delta agonist hydroxy bioisosteres: the discovery of 3-((1-benzylpiperidin-4-yl){4-[(diethylamino)carbonyl]phenyl}amino)benzamide with improved delta agonist activity and in vitro metabolic stability.
Bioorganic & medicinal chemistry letters (2009-10-06)
Andrew M Griffin, William Brown, Christopher Walpole, Martin Coupal, Lynda Adam, Mylene Gosselin, Dominic Salois, Pierre-Emmanuel Morin, Marie Roumi
ABSTRACT
We have investigated phenol replacements in a series of diaryl amino piperidine delta opioid agonists. From this study we have demonstrated that the hydroxy functional group can be replaced with a primary amide group, giving enhanced activity at the delta receptor, increased selectivity versus mu and kappa as well as improved in vitro metabolic stability.