MilliporeSigma
Substituted coumarins as potent 5-lipoxygenase inhibitors.
Bioorganic & medicinal chemistry letters (2006-02-09)
Erich L Grimm, Christine Brideau, Nathalie Chauret, Chi-Chung Chan, Daniel Delorme, Yves Ducharme, Diane Ethier, Jean-Pierre Falgueyret, Richard W Friesen, Jocelyne Guay, Pierre Hamel, Denis Riendeau, Chantal Soucy-Breau, Philip Tagari, Yves Girard
ABSTRACT
Leukotriene biosynthesis inhibitors have potential as therapeutic agents for asthma and inflammatory diseases. A novel series of substituted coumarin derivatives has been synthesized and the structure-activity relationship was evaluated with respect to their ability to inhibit the formation of leukotrienes via the human 5-lipoxygenase enzyme.