MilliporeSigma
  • Synthesis and evaluation of 9-anilinothiazolo[5,4-b]quinoline derivatives as potential antitumorals.
Synthesis and evaluation of 9-anilinothiazolo[5,4-b]quinoline derivatives as potential antitumorals.
European journal of medicinal chemistry (2004-02-28)
Pilar Rodríguez-Loaiza, Angelina Quintero, Rogelio Rodríguez-Sotres, José D Solano, Alfonso Lira-Rocha
ABSTRACT
Five new 9-anilinothiazolo[5,4-b]quinoline derivatives (compounds 5, 7, 9, 10, 11) have been prepared. Some of the compounds were prepared by coupling properly substituted anilines to the novel compound 9-chloro-2-(methylthio)thiazolo[5,4-b]quinoline. Of these, compound 7 (9-anilino-2-[[2-(N,N-diethylamino)ethyl]amino]thiazolo[5,4-b]quinoline) showed the best cytotoxic activity in several cell lines. All compounds demonstrated DNA binding in nanomolar range. Compound 7 inhibited the (14)C-thymidine incorporation into DNA. Results indicate that these derivatives deserve more considerations as potential antitumoral drugs.