MilliporeSigma
  • The effect of N-heterocyclic carbene ligands in the palladium-catalyzed cross-coupling reaction of K [C6F5BF3] with aryl iodides and aryl bromides.
The effect of N-heterocyclic carbene ligands in the palladium-catalyzed cross-coupling reaction of K [C6F5BF3] with aryl iodides and aryl bromides.
Tetrahedron (2008)
Adonin NY, et al.