MilliporeSigma
  • The Competitive Displacement of ?-Lactoglobulin by Tween 20 from Oil-Water and Air-Water Interfaces
The Competitive Displacement of ?-Lactoglobulin by Tween 20 from Oil-Water and Air-Water Interfaces
Journal of Colloid and Interface Science (1993)
Peter J. Wilde, David C. Clark