MilliporeSigma
  • Isolation of lactoferrin from bovine colostrum by ultrafiltration coupled with strong cation exchange chromatography on a production scale
Isolation of lactoferrin from bovine colostrum by ultrafiltration coupled with strong cation exchange chromatography on a production scale
Journal of Membrane Science (2007)
Rong Rong Lu, Shi Ying Xu, Zhang Wang, Rui Jin Yang