MilliporeSigma
  • Comparison of the myoglobin of the zebra (Equus burchelli) with that of the horse (Equus caballus).
Comparison of the myoglobin of the zebra (Equus burchelli) with that of the horse (Equus caballus).
Biochimica et biophysica acta (1975-05-30)
P D Darbre, A E Romero-Herrera, H Lehmann
ABSTRACT
The tryptic and peptic peptides from the myoglobin of the zebra (Equus burchelli) have been compared with those obtained from the myoglobin of the horse (Equus caballus). No differences in the myoglobin were found between these two species.
Sigma-Aldrich
Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder