MilliporeSigma
  • Substrate recognition and channeling of monomodules from the pikromycin polyketide synthase.
Substrate recognition and channeling of monomodules from the pikromycin polyketide synthase.
Journal of the American Chemical Society (2003-10-09)
Brian J Beck, Courtney C Aldrich, Robert A Fecik, Kevin A Reynolds, David H Sherman
ABSTRACT
The unique ability of the pikromycin (Pik) polyketide synthase to generate 12- and 14-membered ring macrolactones presents an opportunity to explore the fundamental processes underlying polyketide synthesis, specifically the mechanistic details of the chain extension process. We have overexpressed and purified PikAIII (module 5) and PikAIV (module 6) and assessed the ability of these proteins to generate tri- and tetraketide lactone products using N-acetylcysteamine-activated diketides and (14)C-methylmalonyl-CoA as substrates. Comparison of the stereochemical specificities for PikAIII and PikAIV and the reported values for the DEBS modules reveals significant differences between these systems.